Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM120B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8356 Supplier Bioss Supplier No. BS8195R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The transcription factor peroxisome proliferator-activated receptor -gamma (PPARg) plays essential roles in adipogenesis by regulating adipocyte-specific genes through association of various co-factors. One such co-factor, PGCC1 (also known as FAM120B), is widely expressed in adult tissues and throughout embryonic development. Overexpression of this protein in OP9 pre-adipocytes promoted their differentiation into adipocytes, and knockdown of PGCC1 expression through RNA interference blocked this process. PGCC1 is homologous to C9orf10 (also known as FAM120A) and has been mapped to chromosome Xp11.22. At least two isoforms of PGCC1 are known to exist.

Specifications

Antigen FAM120B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FAM120B
Gene Accession No. Q96EK7
Gene Alias 4932442K08Rik; AW049283; AW413338; CCPG; Constitutive coactivator of peroxisome proliferator-activated receptor gamma; constitutive coactivator of PPARG; Constitutive coactivator of PPAR-gamma; dJ894D12.1; FAM120B; family with sequence similarity 120, member B; family with sequence similarity 120B; KIAA1838; mKIAA1838; PGCC1; PPARG constitutive coactivator 1; PPARgamma constitutive coactivator 1; Protein FAM120B; RGD1310304
Gene Symbols FAM120B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM120B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84498
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.