Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM129A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555934 Supplier Invitrogen™ Supplier No. PA555934
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILLDETLKVI KEAAILKKHN LFEDNMALPS ESVSSLTDLK PPTGSNQASP ARRASAILPG VLGSETLSNE VFQESEEEKQ PEVPSSLAKG ESLSLPGP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.

Regulates phosphorylation of a number of proteins involved in translation regulation including EIF2A, EIF4EBP1 and RPS6KB1. May be involved in the endoplasmic reticulum stress response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM129A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM129A
Gene Accession No. Q9BZQ8
Gene Alias AI256368; AU019833; C1orf24; Cell growth-inhibiting gene 39 protein; FAM129A; family with sequence similarity 129 member A; family with sequence similarity 129, member A; GIG39; NIBAN; niban apoptosis regulator 1; niban protein; Niban1; Protein FAM129A; Protein Niban; protein Niban 1; protein Niban 1; protein Niban
Gene Symbols FAM129A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM129A. Recombinant protein control fragment (Product #RP-94228).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116496
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.