Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM188B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570401 Supplier Invitrogen™ Supplier No. PA570401
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 77%; Human: 100%; Rabbit: 93%.

MINDY4B (MINDY Family Member 4B) is a Protein Coding gene. An important paralog of this gene is MINDY4. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM188B2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MINDY4B
Gene Accession No. A8MYZ0
Gene Alias C3orf76; CLRN1; FAM188B2; family with sequence similarity 188 member B2; family with sequence similarity 188, member B2; inactive ubiquitin carboxyl-terminal hydrolase MINDY-4B; MINDY family member 4B; MINDY4B; protein FAM188B2; Putative UPF0526 protein B
Gene Symbols MINDY4B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM188B2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 646951
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.