Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM193A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560246 Supplier Invitrogen™ Supplier No. PA560246
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSISTFLGTL ENEHLKKFQV TWELHNKHLF ENLVFSEPLL QSNLPALVSQ IRLGTTTHDT CSEDTYSTLL QRYQRSEEEL RRVAEEWLEC QKRIDAYVDE QM Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

FAM193A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM193A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM193A
Gene Accession No. P78312
Gene Alias C4orf8; Fam193a; family with sequence similarity 193 member A; family with sequence similarity 193, member A; im:7155132; Protein FAM193A; Protein IT14; RES4-22; RGD1309634
Gene Symbols FAM193A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM193A. Recombinant protein control fragment (Product #RP-100143).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8603
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.