Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM26F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116335
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116335 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116335 Supplier Invitrogen™ Supplier No. PA5116335
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FAM26F Polyclonal Antibody for Western Blot

Pore-forming subunit of a voltage-gated ion channel. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM26F
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CALHM6
Gene Accession No. Q5R3K3
Gene Alias C6orf187; Calcium homeostasis modulator protein 6; CALHM6; dJ93H18.5; Fam26f; family with sequence similarity 26 member F; family with sequence similarity 26, member F; LOC100353536; Protein FAM26F; RGD1304835
Gene Symbols CALHM6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 266-295 amino acids from the C-terminal region of human FAM26F.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.