Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM32A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562464 Supplier Invitrogen™ Supplier No. PA562464
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLDKRTPAQA AFEKMQEKRQ MERILKKASK THKQRVEDFN RHLDTLTEHY DIPKVSWTK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM32A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM32A
Gene Accession No. Q9Y421
Gene Alias CGI-144; FAM32A; family with sequence similarity 32 member A; family with sequence similarity 32, member A; OTAG12; OTAG-12; ovarian tumor associated gene-12; ovarian tumor-associated gene 12; Protein FAM32A
Gene Symbols FAM32A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM32A. Recombinant protein control fragment (Product #RP-98968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.