Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542664 Supplier Invitrogen™ Supplier No. PA542664
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DNWVFCGGKG IKTKSPFEQH IKNNKDTNKY EGWPEVVEME GCIPQKQD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 91%; Rat: 100%; Zebrafish: 86%.

FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM3C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM3C
Gene Accession No. Q92520
Gene Alias D6Wsu176e; FAM3 metabolism regulating signaling molecule C; Fam3c; fam3c protein; FAM3C-like protein; family with sequence similarity 3 member C; family with sequence similarity 3, member C; GS3786; Ilei; Interleukin-like EMT inducer; interleukin-like epithelial-mesenchymal transition inducer; predicted osteoblast protein; protein FAM3C; si:dkey-159a18.2; wu:fb99h11; wu:fi35h03; zgc:55444; zgc:77090
Gene Symbols FAM3C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FAM3C (aa 180-227).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10447
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.