Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM46C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107035 Supplier Thermo Scientific Supplier No. PA5107035
Only null left
Add to Cart
Add to Cart

FAM46C Polyclonal Antibody

FAM46C Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM46C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene FAM46C
Gene Accession No. Q5VWP2
Gene Alias 4930431B09Rik; AI449797; DN-384L23.3; Fam46c; family with sequence similarity 46 member C; family with sequence similarity 46, member C; Non-canonical poly(A) polymerase FAM46C; protein FAM46C; Protein FAM46C-like protein; putative nucleotidyltransferase FAM46C; TENT5C; Terminal nucleotidyltransferase 5C
Gene Symbols FAM46C
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: SETIPIHGRG NFPTLELQPS LIVKVVRRRL AEKRIGVRDV RLNGSAASHV.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54855
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.