Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM50B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578468 Supplier Invitrogen™ Supplier No. PA578468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, MCF-7, MDA-MB-231 Predicted Reactivity: Mouse (89%), Rat (90%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FAM50B contains an intronless ORF that arose from ancestral retroposition. The encoded protein is related to a plant protein that plays a role in the circadian clock. FAM50B is adjacent to a differentially methylated region (DMR) and is imprinted and paternally expressed in many tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM50B
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene FAM50B
Gene Accession No. Q9Y247
Gene Alias D0H6S2654E; D6S2654E; FAM50B; family with sequence similarity 50 member B; family with sequence similarity 50, member B; protein FAM50B; Protein XAP-5-like; RGD1563458; X5l; XAP-5-like; XAP5-like protein
Gene Symbols FAM50B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FAM50B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26240
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.