Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM54A Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
SDP

Catalog No. 501684027
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4027 Supplier OriGene Supplier No. TA811420
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM54A belongs to the MTFR1/FAM54 family. The function of the FAM54A protein is not known.

Specifications

Antigen FAM54A
Applications Western Blot
Classification Monoclonal
Clone OTI2G8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MTFR2
Gene Accession No. B0K035, Q6P444
Gene Alias DUFD1, FAM54A
Gene Symbols Mtfr2
Host Species Mouse
Immunogen Full length human recombit protein of human FAM54A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100911069, 113115, 483997
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.