Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM71B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569849 Supplier Invitrogen™ Supplier No. PA569849
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Human: 100%.

FAM71B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM71B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM71B
Gene Accession No. Q8TC56
Gene Alias FAM71B; family with sequence similarity 71 member B; family with sequence similarity 71, member B; HSD51; HSD-51; Protein FAM71B; testicular tissue protein Li 68; Unknown (protein for IMAGE:8061995)
Gene Symbols FAM71B
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FAM71B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153745
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.