Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546441 Supplier Invitrogen™ Supplier No. PA546441
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ANGRLSSSSG SASDRTSSNP FSGRSAGSHP VPESKQNKTK TKKQTTLWFL Sequence homology: Human: 100%.

Probable proto-oncogene that functions in the epidermal growth factor receptor/EGFR signaling pathway. Activates both RAS/MAPK and PI3K/AKT/TOR signaling cascades downstream of EGFR. Required for the RAS/MAPK signaling cascade activation upon EGFR stimulation, it also activates both signaling cascades independently of EGFR activation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FAM83A
Gene Accession No. Q86UY5
Gene Alias BJ-TSA-9; FAM83A; family with sequence similarity 83 member A; MGC14128; protein FAM83A; TSGP; Tumor antigen BJ-TSA-9; tumor-specific gene expressed in prostate protein
Gene Symbols FAM83A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FAM83A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84985
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.