Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FAM83H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555121 Supplier Invitrogen™ Supplier No. PA555121
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVSQAWREEV AAPGAVGGER RSLESCLLDL RDSFAQQLHQ EAERQPGAAS LTAAQLLDTL GRSGSDRLPS RFLSAQSHST SPQGLDSPLP LEGS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

The protein encoded by this gene plays an important role in the structural development and calcification of tooth enamel. Defects in this gene are a cause of amelogenesis imperfecta type 3 (AI3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAM83H
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM83H
Gene Accession No. Q6ZRV2
Gene Alias AA409316; AI3; FAM83H; FAM83H variant 1; FAM83H variant 2; family with sequence similarity 83 member H; family with sequence similarity 83, member H; FLJ46072; protein FAM83H; RGD1305866
Gene Symbols FAM83H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FAM83H. Recombinant protein control fragment (Product #RP-93934).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.