Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM84B Monoclonal Antibody (OTI5G1), TrueMAB™, OriGene
SDP

Catalog No. p-200010357
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1885 30 μL
50-167-3064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1885 Supplier OriGene Supplier No. TA501969S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FAM84B Monoclonal antibody specifically detects FAM84B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen FAM84B
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI5G1
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FAM84B
Gene Accession No. Q96KN1
Gene Alias BCMP101; BF642811; breast cancer membrane protein 101; breast cancer membrane-associated protein 101; D330050I23Rik; FAM84B; family with sequence similarity 84 member B; family with sequence similarity 84, member B; LRAT domain-containing 2; LRATD2; neurological/sensory 2; NSE2; Protein FAM84B; Protein LRATD2; Protein NSE2; RGD1562236
Host Species Mouse
Immunogen Full length human recombinant protein of human FAM84B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157638
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.