Learn More
AdipoGen FAMC
SDP
Supplier:  AdipoGen CDXD0099M050
Encompass_Preferred
Chemical. CAS 127192-67-0. Formula C13H8F2N4O3. MW 203.28. Synthetic. Polarity fluorescent probe.
Catalog No. 50-256-1240
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.