Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Fan Cart
SDPSCI_ED

Catalog No. S07388 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity guide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S07388 Activity guide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07388 Supplier United Scientific Supplies Supplier No. FNCRT1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrate Newton's principle of action and reaction using this basic and inexpensive fan cart.

  • Powered by two AA batteries (not included) contained in a removable power unit
  • Controlled by a switch
  • Propeller delivers thrust of about 88mN and accelerates cart at 0.4m/s2
  • Attach a transparent plate to show effect of reversing airflow
  • Includes activity guide

Specifications

Product Type Fan Cart
For Use With (Application) Demonstrate Newton's principle of action and reaction
Includes Activity guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.