Learn More
Fan Cart
SDPSCI_ED
Catalog No. S52137 Shop All Science First Products
Change view
Click to view available options
Fan cart
Catalog No.
S52137 Fan cart
1 options
Catalog No. S52137 Supplier Science First Supplier No. 10105
Requires
Four AA batteries
Demonstrates acceleration.
  • Self-propelled fan cart
  • Without the sail in place, demonstrates uniform acceleration independent of gravity
  • Study constant or uniform acceleration
  • Velocity of the cart is a function of the sail's angle
Specifications
Product Type Fan Cart
For Use With (Application) Studying motion with a constant force, exploring Newton's third law or forces at an angle