Learn More
StatLab FAN PAD GL LARGE 8X25IN 30CS
SDP
Supplier:  StatLab FAN825
Encompass_Preferred
Formaldehyde neutralizing pads measure 8 in. and 25 in..
Catalog No. NC9102526
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.