Learn More
IMEB, Inc Fan (Formaldehyde Absorbent Neutralizer) Pads Mini, 6 Rolls per Case
Supplier: IMEB, Inc FAN55
These pads are specially treated to absorb formalin and formaldehyde. Formalin products are absorbed and harmful vapors neutralized. Pads can be disposed in regular trash receptacle. 11" X 10 3/4", 55 per roll, 6 rolls per case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.