Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FANCA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596755 Supplier Invitrogen™ Supplier No. PA596755
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSDSWVPNSA SGQDPGGRRR AWAELLAGRV KREKYNPERA QKLKESAVRL LRSHQDLNAL LLEVEGPLCK KLSLSKVIDC DSSEAYANHS SSFIGSALQD QASRLGVPVG ILSAGMVASS VGQICTAPAE TSHPVLLTVE QRKKLSSLLE FAQYLLAHSM FSRLSFCQEL WKIQSSLLLE AVWHLHVQGI VSLQELLESH PDMHAVGSWL FRNLCCLCEQ MEASCQHADV ARAMLSDFVQ MFVLRGFQKN SDLRRTVEPE KMPQVTVDVL QRMLI; Positive Samples: HeLa; Cellular Location: Cytoplasm, Nucleus.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that are known calcium sensors and mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. The encoded protein is also a substrate for ubiquitin-E3-ligase parkin. The gene has previously been referred to as synaptotagmin XII but has been renamed synaptotagmin XI to be consistent with mouse and rat official nomenclature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FANCA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FANCA
Gene Accession No. O15360
Gene Alias AW208693; FA; FA1; FAA; FACA; FAH; FA-H; Fanca; FANCH; Fanconi anemia complementation group A; Fanconi anemia group A protein; Fanconi anemia group A protein homolog; Fanconi anemia, complementation group A; Fanconi anemia, complementation group H; Fanconi anemia, type 1; Protein FACA
Gene Symbols FANCA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-275 of human FANCA (NP_0001262).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2175
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.