Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FANCD2 Polyclonal Antibody, HRP
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522707
Description
In Western blot, this antibody should recognize a band at ~166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights.
FANCD2 (Fanconi anemia group D2 protein) is required for maintenance of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.
Specifications
FANCD2 | |
Polyclonal | |
HRP | |
FANCD2 | |
2410150O07Rik; AU015151; BB137857; DKFZp762A223; FA4; FACD; FACD2; FAD; FAD2; FA-D2; FANCD; FANCD2; Fanconi anemia complementation group D2; Fanconi anemia D2 protein; Fanconi anemia group D2 protein; Fanconi anemia group D2 protein homolog; Fanconi anemia, complementation group D2; FLJ23826; Protein FACD2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
211651, 2177, 312641, 394241, 484659 | |
4°C, store in dark | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.82 mg/mL | |
PBS with no preservative | |
Q6IV68, Q80V62, Q9BXW9 | |
FANCD2 | |
Human FANCD2 fusion protein (N-terminal fragment). | |
50 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Avian, Zebrafish, Marsupial | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction