Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Readers to Eaters Farmer Will Allen and the Growing Table
SCI_ED

Catalog No. S000947 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S000947 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000947 Supplier Readers to Eaters Supplier No. 9780983661580
Add to Cart
Add to Cart

Educational book by Jacqueline Briggs Martin about inspiring food heroes.

  • First book in the Food Heroes series that highlights inspiring stories about food and community

Specifications

Grade K to 2
Class Size 1 Student
Product Type Farmer Will Allen and the Growing Table
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.