Learn More
Invitrogen™ Farnesyl Polyclonal Antibody

Description
Farnesyl Polyclonal Antibody for ICC/IF, ELISA
Specifications
Specifications
| Antigen | Farnesyl |
| Applications | ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Host Species | Rabbit |
| Immunogen | Farnesyl cysteine conjugated to KLH. |
| Purification Method | Ammonium Sulfate Precipitation |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.