Learn More
Enzo Life Sciences Fas monoclonal antibody (ZB4) (100 µg)
Supplier: Enzo Life Sciences ADIAAM227E
Clone: ZB4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Fas. UniProt: P25445. Species Reactivity: Human. Applications: Flow Cytometry, WB. FUNC. Recommended dilutions: Flow Cytometry (5-20µg/ml). Western Blot (5µg/ml, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.