Learn More
MSA™ Fas-Trac™ III Replacement Suspension
Catalog No. 19141451 Shop All MSA Products
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-141-451 Large
1 options
Catalog No. 19-141-451 Supplier MSA™ Supplier No. 10148707
Add to Cart
Add to Cart
MSA™ Fas-Trac™ III Replacement Suspension reduces pressure headaches, pulled hair, and the need to sacrifice worker comfort for safety.
  • Designed with input from hundreds of customers
  • For use with MSA Helmets, as specified
  • Fas-Trac™ III ratchet suspension (4-point and 6-point, as noted)
  • Flush rear lug attachments eliminate pressure points
  • Three levels of nape strap adjustment for customized fit
  • Graspable ratchet with ultra-smooth rotation and steadfast hold
  • The ratchet comfort pad contours to the back of the head to improve airflow and prevent hair-pulling
Specifications
Certifications/Compliance ELECTROSTATIC-INERIS
Product Type Helmet accessory
Quantity 1 Ea.
Size Large
Weight (English) 0.216 lb
Weight (Metric) 0.098 kg