Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA V-Gard C1™ Full Brim Fas-Trac III Vented Hard Hat

Catalog No. 19815756 Shop All MSA Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-815-756 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-815-756 Supplier MSA V-Gard C1™ Full Brim Supplier No. 10215828
Add to Cart
Add to Cart
This item is not returnable. View return policy

Patent pending ReflectIR™ Thermal Barrier keeps the inside of the hard hat up to 20°F (11°C) cooler in sunny conditions. Footnote Required: Compared to a hard hat without the ReflectIR thermal barrier; testing done with black, non-vented, V-Gard C1™ hard hats.

  • The V-Gard C1™ Hard Hat is a general purpose full brim hard hat, particularly suited for outdoor workers in sunny conditions.
  • The patent pending ReflectIR™ thermal barrier keeps the inside of the hard hat up to 20°F (11°C) cooler in sunny conditions, while a moisture-wicking padded sweatband keeps you drier and more comfortable.
  • The brim and optional vents are the largest offered on a full brim V-Gard to provide additional shade and significant air flow.
  • The V-Gard C1 Hard Hat is compatible with all V-Gard full brim accessories for superior versatility and ease of use.

Specifications

Certifications/Compliance ANSI/ISEA Z89.1-2014 (R2019), Type I, Class C, LT (Low Temp), HT (High Temp), Reverse Donning; CSA Z94.1-15, Type I, Class C
Color White
For Use With (Equipment) V-Gard Hard Hat Accessories
Product Type Hard Hat
Size Standard (Medium)
Ventilated Ventilated
Head Style Full Brim
Width (Metric) 279 mm
Width (English) 10.98 in.

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.