Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fascin Mouse anti-Human, Clone: 1B5G3, Proteintech
SDP

Catalog No. 501736753 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6753 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6753 Supplier Proteintech Group Inc Supplier No. 663211IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Specifications

Antigen Fascin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B5G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FSCN1
Gene Accession No. Q16658
Gene Alias 55 kDa actin bundling protein, FAN1, Fascin, Fascin 1,Fascin, FSCN1, HSN, p55, Singed like protein, SNL
Gene Symbols FSCN1
Host Species Mouse
Immunogen Fascin Fusion Protein Ag6074
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6624
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.