Learn More
ACROBiosystems Recombinant Protein;Human Fas Ligand / TNFSF6 Protein;HEK293;1MG;FAL,FASLG,ALPS1B,APT1LG1,CD178,CD95-L,CD95L,FASL,TNFSF6,Fas ligand
Supplier:  ACROBiosystems FALH52411MG
Encompass_Preferred
Recombinant Protein;Human Fas Ligand / TNFSF6 Protein;1MG;Human Fas Ligand, His Tag (FAL-H5241) is expressed from human 293 cells (HEK293). It contains AA Pro 134 - Leu 281 (Accession # AAH17502.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa. The protein migrates as 25-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Fas Ligand, His Tag (Cat. No. FAL-H5241) at 5μg/mL (100 μL/well) can bind Human Fas, Fc Tag (Cat. No. FAS-H5252) with a linear range of 1.56-12.5 ng/mL (QC tested).
Catalog No. 50-165-7583
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.