Learn More
Creative Biomart Insulin Aspart
Supplier:  Creative Biomart THP0124
Encompass_Preferred
The product is a recombinant, biosynthetic, fast-acting insulin analogue. It has a single amino acid substitution at position B28 where proline is replaced with aspartic acid. This substitution decreases its propensity to form hexamers. The product is produced in a genetically modified strain of Saccharomyces cerevisiae and harvested from a bioreactor.
Catalog No. 50-208-4175