Learn More
Abcam Fast Bisulfite Conversion Kit, 30 min, 50 Test
Description
- Fast bisulfite conversion in 30 min
- Cited in 4 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | DNA (Methylated) |
| Substrate | DNA |
| Run Time | 30 min. |
| Format | Kit |
| No. of Reactions | 50 Tests |
| Quantity | 50 Tests |
| For Use With (Application) | DNA methylation analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.