Learn More
New England Biolabs, Inc. Fast DNA Ladder – 1 ml
SDP
Supplier:  New England Biolabs, Inc. N3238S
Encompass
The Fast DNA ladder is a pre-mixed, ready-to-load molecular weight marker containing xylene cyanol FF dye as a tracking dye. The DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 11 bands suitable for use as molecular weight standards for fast electrophoresis systems as well as standard electrophoresis. The digested DNA includes fragments ranging from 50 base pairs to 10 kilobases. The 1 kb fragment has increased intensity to serve as reference band. This DNA Ladder has been tested and is suitable for use with many fast electrophoresis systems, such as the FlashGel DNA System from Lonza or the E-Gel Precast Agarose Electrophoresis System from Invitrogen.
Catalog No. NC9015664