Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Garnet GBC Base 97%, Thermo Scientific Chemicals

Catalog No. AC153261000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153261000 100 g
AC153260250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153261000 Supplier Thermo Scientific Chemicals Supplier No. 153261000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-56-3
Molecular Formula C14H15N3
Molecular Weight (g/mol) 225.295
MDL Number MFCD00007733
InChI Key PFRYFZZSECNQOL-UHFFFAOYSA-N
Synonym 2-Aminoazotoluene, 4'-Amino-2, 3'-dimethylazobenzene, C.I. 11160, Solvent Yellow 3
PubChem CID 7340
IUPAC Name 2-methyl-4-[(2-methylphenyl)diazenyl]aniline
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)N)C

Specifications

Assay Percent Range 97%
Linear Formula NH2C6H3(CH3)N=NC6H4CH3
Beilstein 16, 344
Merck Index 15, 415
Infrared Spectrum Authentic
Formula Weight 225.29
Percent Purity ≥96.0%
UV Spectrometry Complies
Color Brown/Red
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Fast Garnet GBC base, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
May cause cancer.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-591-2
RTECSNumber XU8800000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.