Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Garnet GBC Salt 90%, Thermo Scientific™

Catalog No. AC191330250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC191330250 25 g
AC191331000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191330250 Supplier Thermo Scientific Chemicals Supplier No. 191330250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-89-3
Molecular Formula C14H14N4O4S
Molecular Weight (g/mol) 334.35
InChI Key PPTXFSQSISUDAU-UHFFFAOYSA-M
Synonym 2-Methyl-4-(2-methylphenylazo)benzenediazonium sulfate, Azoic Diazo No. 4, C.I. 37210
PubChem CID 66872
IUPAC Name hydrogen sulfate;2-methyl-4-[(2-methylphenyl)diazenyl]benzenediazonium
SMILES CC1=CC=CC=C1N=NC2=CC(=C(C=C2)[N+]#N)C.OS(=O)(=O)[O-]

Specifications

Infrared Spectrum Authentic
Formula Weight 334.34
Percent Purity ≥88%
UV Spectrometry Complies
Color Brown to Brown/Rust
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fast Garnet GBC salt, 90%
EINECSNumber 202-985-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.