Learn More
Abcam Fast Green FCF, Tissue and Protein Stain, 25 g
Description
- Molecular Weight: 808.86 g/mol
- Form: Solid
- Applications: Tissue and protein stain, quantitative stain for histones at alkaline pH, protein stain in IEF and SDS-PAGE electrophoresis, food dye
- Features: Fluoresces in near-infrared (NIR), replacement for Light Green SF yellowish in Masson's trichrome
- Composition: 100% Fast green FCF
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Proteins, Histones |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Form | Solid |
| Detection Method | Fluorescence |
| Buffer | 100% Fast green FCF |
| Content And Storage | Store at ambient temperature. |
| Detection Location | In-Gel Detection |
| Formulation | 100% Fast green FCF |
| Quantity | 25 g |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.