Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ FastDigest SmaI

Catalog No. p-200089022
Change view
Click to view available options
Quantity:
100 μL (100 Reactions)
200 μL (200 Reactions)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERFD0663 100 μL (100 Reactions)
FERFD0664 200 μL (200 Reactions)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERFD0663 Supplier Thermo Scientific™ Supplier No. FD0663
Only null left
Add to Cart
Edge
Add to Cart

The FastDigest SmaI restriction enzyme recognizes CCC^GGG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (isoschizomers: TspMI, XmaCI; neoschizomer: XmaI).

5' CCC ↓ GGG 3'

3' GGG ↑ CCC 5'

FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double- or multiple-DNA digestion within 5-15 minutes, eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA ligase, alkaline phosphatases, and T4 DNA polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

  • 100% activity of all FastDigest enzymes in the universal buffer
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5–15 minutes
  • Direct loading on gels
  • No star activity
  • 176 FastDigest enzymes available

Applications

  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP analysis

Notes

  • The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer.
  • For applications that require product analysis by fluorescence excitation (e.g., concentration measurements in UV light), the colorless FastDigest Buffer is recommended.
  • For methylation sensitivity, refer to product specifications.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage FastDigest SmaI (100 μL), 10X FastDigest Buffer (1.0 mL), 10X FastDigest Green Buffer (1.0 mL), Store at –20°C.
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Sma I
Compatible Buffer 10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 100 μL (100 Reactions)
Product Type Restriction Enzyme
Research Category Traditional Cloning
Product Line FastDigest
Overhang Blunt
Inactivation Temperature 65°C
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.