Learn More
Thomas Scientific FastPette Pro Pipet Controller with two-position charging stand, additional autoclavable filter 0.2æm, and universal power supply (suitable for US, EU, UK and AU)
Supplier: Thomas Scientific 1185L54
The new FASTPETTE™ PRO- Lightweight and ergonomically shaped motorized pipette controller designed for work with glass or plastic serological pipettes from 0.5 to 100 ml range. Conveniently positioned switches allow choosing different operation modes and speed depending on the volume of the serological pipette and liquid viscosity. Additionally aspirating and dispensing speed is controlled through two comfortable finger triggers. LCD Display Pipette When fully charged, the NiMH batteries enable up to eight hours of continuous use. The large LCD display the serological high viscosity faspette indicates the battery status along with the pipetting mode and speed selected. In order to protect the device against overfilling the FASTPETTE™ PRO is equipped with a 0.2 um PTFE filter. The filter, the pipette holder and the nosepiece are autoclavable. The FASTPETTE™ PRO is also supplied with a universal battery charger, an additional hydrophobic 0.2 µm filter and a two-po
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.