Learn More
JDM SCIENTIFIC LLC FASTWORX-S SORBENT POWDER - 10
Supplier:  JDM SCIENTIFIC LLC FASTWORXS1004
Encompass_Preferred
Faster Chemistrys FastWoRX-S is a hydrophobic powder that absorbs most organics. It can easily and quickly be separated along with the absorbed organics
Catalog No. NC2127766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.