Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fat Brown RR, MP Biomedicals™

Catalog No. ICN15803525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15803525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15803525 Supplier MP Biomedicals, Inc Supplier No. 0215803525

Chemical Identifiers

CAS 6416-57-5
Molecular Formula C16H14N4
Molecular Weight (g/mol) 262.32
MDL Number MFCD00009689
InChI Key AHWMWMNEYBHQNL-UHFFFAOYSA-N
Synonym Solvent brown 1, C.I. 11285
PubChem CID 62608
IUPAC Name 4-[2-(naphthalen-1-yl)diazen-1-yl]benzene-1,3-diamine
SMILES NC1=CC=C(N=NC2=C3C=CC=CC3=CC=C2)C(N)=C1

Specifications

Formula Weight 262.31
Color Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fat Brown RR
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.