Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FABP1 (L-FABP) Recombinant Protein

Catalog No. PIRP75549
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75549 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75549 Supplier Invitrogen™ Supplier No. RP75549
Add to Cart
Add to Cart

Recombinant Protein

This product contains a purified 127 amino acid fragment (aa1-127) of human FABP-L protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07148
Concentration 0.745 mg/mL
For Use With (Application) Control, Western Blot
Formulation tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 2168
Molecular Weight (g/mol) 18.71 kDa
Name Human FABP1 (L-FABP)
Quantity 100 μg
Source E. Coli
Regulatory Status RUO
Gene Alias 14 kDa selenium-binding protein; FABP1; FABPL; Fabplg; fatty acid binding protein 1; fatty acid binding protein 1, liver; fatty acid binding protein liver; fatty acid-binding protein 1; Fatty acid-binding protein, liver; L-FABP; Liver-type fatty acid-binding protein; p14; SCP; SCP.; Squalene- and sterol-carrier protein; Z-protein
Common Name FABP1
Gene Symbol FABP1
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1-Ile127 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.