Learn More
Medchemexpress LLC FAUC 365 10mg | 474432-66-1 | 462.44 g/mol | C23H25Cl2N3OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11602010MG
Encompass_Preferred
FAUC 365 is a small-molecule dopamine D3 receptor-selective antagonist used in neuropharmacology research. It shows high affinity for the D3 receptor (Ki ≈ 0.5 nM) and is supplied as a high-purity powder for in vitro and in vivo studies.
  • High affinity for dopamine D3 receptor (Ki ≈ 0.5 nM).
  • Demonstrates selectivity over D2 and D4 receptor subtypes.
  • High purity suitable for research applications (purity reported ~98.75%).
  • Available in small pack sizes, including 10 mg.
  • Soluble in DMSO; in vivo formulation protocols provided.
  • Store powder at recommended temperatures to maintain stability.
Catalog No. 50-002-29799
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.