Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599815 Supplier Invitrogen™ Supplier No. PA599815
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FBF1.

Keratin-binding protein required for epithelial cell polarization. Involved in apical junction complex (AJC) assembly via its interaction with PARD3. Required for ciliogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FBF1
Gene Accession No. A2A870, Q8TES7
Gene Alias 1110033G01Rik; 5930413D20; Alb; albatross; Fas (TNFRSF6) binding factor 1; Fas binding factor 1; Fas binding protein 1; fas-binding factor 1; Fbf1; FBF-1; KIAA1863; likely ortholog of H. sapiens Fas (TNFRSF6) binding factor 1 (FBF1); mKIAA1863; Protein albatross
Gene Symbols FBF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FBF1(Accession Q8TES7), corresponding to amino acid residues E753-E803.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217335, 85302
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.