Learn More
Thomas Scientific Fetal Bovine Serum (FBS) - Premium US Source, 500mL
SDP
Supplier:  Thomas Scientific C788U20
Encompass
High quality FBS at an affordable price From USDA licensed facilities inside and outside the U.S. Triple 0.1Ým filtered Endotoxin < 2.0 EU/mL Tested for Growth Promotion* Large lot sizes
Catalog No. NC0953215