Learn More
Thomas Scientific FBS USDA Sourced | Heat Inactivated | 500ml
SDP
Supplier:  Thomas Scientific C788U23
Encompass_Preferred
FBS USDA Sourced, Heat Inactivated is virus screened, mycoplasma tested and triple 0.1 micron filtered. It should be stored at -20 degrees celcius and is shipped on dry ice.
Catalog No. NC1857274