Learn More
FBW7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301720AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Cyclin E, one of the activators of the cyclin-dependent kinase Cdk2, is critical for the initiation of DNA replication and other S-phase functions. The proper timing and amplitude of cyclin E expression seem to be important, because elevated levels of cyclin E have been associated with a variety of maligcies and constitutive expression of cyclin E leads to genomic instability. Phosphorylated cyclin E depends on the novel human gene, hCdc4, that was found to be mutated in a cell line derived from breast cancer that expressed extremely high levels of cyclin E.Specifications
| FBW7 | |
| Polyclonal | |
| Unconjugated | |
| FBXW7 | |
| AGO, archipelago, Archipelago homolog, archipelago homolog (Drosophila), CDC4, F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, FBW6, FBW7, FBX30, FBXO30, FBXW6, FLJ11071, hAgo, hCdc4, homolog of C elegans sel-10, SEL10, SEL-10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55294 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q969H0 | |
| FBXW7 | |
| Between 1 and 50 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.