Learn More
FBW7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Specifications
Specifications
| Antigen | FBW7 |
| Applications | Immunohistochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | FBXW7 |
| Gene Accession No. | Q969H0 |
| Gene Alias | AGO, archipelago, Archipelago homolog, archipelago homolog (Drosophila), CDC4, F-box and WD repeat domain containing 7, E3 ubiquitin protein ligase, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, FBW6, FBW7, FBX30, FBXO30, FBXW6, FLJ11071, hAgo, hCdc4, homolog of C elegans sel-10, SEL10, SEL-10 |
| Gene Symbols | FBXW7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.