Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL10 Monoclonal Antibody (3B7D11)
GREENER_CHOICE

Catalog No. PIMA531731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531731 Supplier Invitrogen™ Supplier No. MA531731
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31731 has been tested in indirect ELISA.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs, which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL10
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3B7D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene KDM2B
Gene Accession No. Q8NHM5
Gene Alias [Histone-H3]-lysine-36 demethylase 1B; Cxxc2; CXXC-type zinc finger protein 2; Fbl10; F-box and leucine-rich repeat protein 10; F-box protein containing leucine repeats 10; F-box protein FBL10; F-box/LRR-repeat protein 10; Fbxl10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; Jhdm1b; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; Kdm2b; Kiaa3014; lysine (K)-specific demethylase 2B; lysine demethylase 2B; lysine-specific demethylase 2B; NDY1; PCCX2; Protein JEMMA; Protein-containing CXXC domain 2
Gene Symbols KDM2B
Host Species Mouse
Immunogen Purified recombinant fragment of human FBXL10 (AA: 457-555) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.