Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FBXL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564927 Supplier Invitrogen™ Supplier No. PA564927
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INRNSIPLQLP VEALQKGCPQ LQVLRLLNLM WLPKPPGRGV APGPGFPSLE ELCLASSTCN FVSNEVLGRL LHGSPNLRLL DLRGCARITP AGLQDLPCRE LEQLHLGLYG TSD Highest antigen sequence identity to the following orthologs - mouse 83%, rat 82%.

Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBXL6
Gene Accession No. Q8N531
Gene Alias FBL6; FBL6A; F-box and leucine rich repeat protein 6; F-box and leucine-rich repeat protein 6; F-box protein FBL6; F-box/LRR-repeat protein 6; FBXL6; PP14630
Gene Symbols FBXL6
Host Species Rabbit
Immunogen Recombinant Human FBXL6. Recombinant protein control fragment (Product #RP-104557).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.