Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXO31 Antibody (OTI1H1) Alexa Fluor™ Plus 594, Novus Biologicals™
SDP

Catalog No. RD10121262
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10121262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10121262 Supplier R&D Systems Supplier No. NBP272208AFP594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

FBXO31 Monoclonal antibody specifically detects FBXO31 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence

Specifications

Antigen FBXO31
Applications Flow Cytometry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI1H1
Conjugate Alexa Fluor Plus 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp434B027, F-box only protein 31, F-box protein 31, FBX14DKFZp434J1815, Fbx31, FBXO14FLJ22477, MGC15419, MGC9527, pp2386, putative breast cancer tumor-suppressor, SCF ubiquitin ligase specificity factor
Host Species Mouse
Immunogen Full length human recombinant protein of human FBXO31(NP_079011) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79791
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.