Learn More
Invitrogen™ FBXO7 Polyclonal Antibody

Description
Immunogen sequence: PTLGHLRSHLR QSLLCTWGYS SNTRFTITLN YKDPLTGDEE TLASYGIVSG DLICLILQDD IPAPNIPSST DSEHSSLQNN EQPSLA Highest antigen sequence identity to the following orthologs - mouse 79%, rat 79%.
Specifications
Specifications
| Antigen | FBXO7 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FBXO7 |
| Gene Accession No. | Q9Y3I1 |
| Gene Alias | 2410015K21Rik; A230052G17Rik; AI117679; AW260462; C85118; DKFZp686B08113; F-box only protein 7; F-box protein 7; FBX; FBX07; FBX7; FBXO7; hypothetical protein LOC558097; PARK15; PKPS; si:ch211-286m4.2; Unknown (protein for MGC:134509); zgc:114096 |
| Gene Symbols | FBXO7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.