Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fc Block , Unlabeled, Clone: 3070, BD

Catalog No. BDB564219 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564219 50 μg
BDB564220 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB564219 Supplier BD Biosciences Supplier No. 564219
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monoclonal Antibody

FcγReceptors belong to the immunoglobulin superfamily and are expressed at varying levels in multiple cell lineages including high expression in myeloid and B cells. The major functions of Fc receptors are protective functions of the immune system. There are multiple different types of Fc receptors reflecting a variety of different biological activities, which are modulated when they are aggregated by multivalent antigen-antibody complexes.

While normally serving important physiological roles in the immune system, Fc Receptors can also be the cause of nonspecific, false-positive antibody staining of cells. Human BD Fc Block™ is designed and formulated to block or significantly reduce potential non-specific antibody staining caused by receptors for IgG that may be encountered in various applications including the flow cytometric analysis of human cells. Moreover, it can increase the specificity of antibody labeling of extremely rare target cells such as antigen-specific B cells, fetal cells in maternal blood, hematopoietic progenitor cells, or disseminated epithelial tumor cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc Block
Applications Flow Cytometry
Classification Monoclonal
Clone 3070
Concentration 0.5mg/mL
Conjugate Unconjugated
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Product Type Antibody
Form Purified
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.